missing translation for 'onlineSavingsMsg'
Learn More
2-n-Butoxyethanol, 99%, Thermo Scientific Chemicals Product Code.: 11499106

2-n-Butoxyethanol, 99%, Thermo Scientific Chemicals

Product Code. 11499106
500 mL
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
This item is not returnable. View return policy

Product Code. 11499106

Brand: Thermo Scientific Alfa Aesar A17976.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in industry for degreasing and detergent applications.2-n-Butoxyethanol is used in the industry for degreasing and detergent applications. It acts as a surfactant in domestic and industrial products. It is an active component of fracturing fluids, drilling stabilizers and oil slick dispersants. It is also used as a solvent for paints, surface coatings, cleaning products and inks. Further, it is employed as a food additive, antimicrobial agent, defoamer, stabilizer and adhesive. In addition to this, it is used as a whiteboard cleaner, liquid soap, cosmetic, dry cleaning solution, lacquer, varnishe and latex paint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in industry for degreasing and detergent applications.2-n-Butoxyethanol is used in the industry for degreasing and detergent applications. It acts as a surfactant in domestic and industrial products. It is an active component of fracturing fluids, drilling stabilizers and oil slick dispersants. It is also used as a solvent for paints, surface coatings, cleaning products and inks. Further, it is employed as a food additive, antimicrobial agent, defoamer, stabilizer and adhesive. In addition to this, it is used as a whiteboard cleaner, liquid soap, cosmetic, dry cleaning solution, lacquer, varnishe and latex paint.

Solubility
Miscible with water, ethyl alcohol, ethyl ether, acetone, n-heptane benzene, mineral oil, aromatic paraffin and halogenated hydrocarbons. Slightly miscible with carbon tetrachloride.

Notes
Excellent Hydrophilic-lipophilic balance (HLB).Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, metals, aluminum and alkalies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00002884
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether ebShow MoreShow Less
PubChem CID 8133
ChEBI CHEBI:63921
IUPAC Name 2-butoxyethanol
SMILES CCCCOCCO

Specifications

Melting Point °C to 79°C
Density 0.902
Boiling Point 171°C to 173°C
Flash Point 64°C (147°F)
Assay Percent Range 0.98
Linear Formula CH3(CH2)3OCH2CH2OH
Refractive Index 1.419
Quantity 500 mL
UN Number UN2810
Beilstein 1732511
Merck Index 14,1559
Solubility Information Miscible with water,ethyl alcohol,ethyl ether,acetone,n-heptane benzene,mineral oil,aromatic paraffin and halogenated hydrocarbons. Slightly miscible with carbon tetrachloride.
Formula Weight 118.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-n-Butoxyethanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253268 Certificate TypeCertificate of Analysis Date28/11/2024 Catalog NumberA17976.0D,A17976.0F,A17976.AP
Lot Number10253269 Certificate TypeCertificate of Analysis Date28/11/2024 Catalog NumberA17976.0D,A17976.0F,A17976.AP
Lot Number10240378 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog NumberA17976.0D,A17976.0F,A17976.AP
Lot Number10238178 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17976.0D,A17976.0F,A17976.AP
Lot Number10236258 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17976.0D,A17976.0F,A17976.AP
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-n-Butoxyethanol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.