missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Butyl Ether, ≥99%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Specifications
Specifications
CAS | 111-76-2 |
Melting Point | -74.8°C |
Color | Colorless |
Density | 0.902g/mL at 25°C |
Boiling Point | 168°C to 172°C (1013 hPa) |
Explosion Limit | 0.106 |
Flash Point | 67°C (152.6°F) |
Ignition Point | 473°F |
Odor | Odorless |
Packaging | Glass bottle |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Safety and Handling

Product Identifier
- Ethylene glycol butyl ether
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 2-butoxyethanol; ethylene glycol monobutyl ether; butyl cellosolve
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethylene Glycol Butyl Ether, ≥99%, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction