missing translation for 'onlineSavingsMsg'
Learn More
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%, Thermo Scientific Chemicals Product Code.: 15438019

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%, Thermo Scientific Chemicals

Product Code. 15438019
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15438019

Brand: Thermo Scientific Alfa Aesar J66601.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

Solubility
Soluble in water. (879 g/L) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129499-78-1
Molecular Formula C12H18O11
Molecular Weight (g/mol) 338.265
MDL Number MFCD23701380
InChI Key MLSJBGYKDYSOAE-DCWMUDTNSA-N
Synonym l-ascorbic acid 2-glucoside, aa-2g, unii-2v52r0nhxw, 2-o-alpha-d-glucopyranosyl-l-ascorbic acid, 2v52r0nhxw, l-ascorbic acid-2-glucoside, ascorbic acid 2-o-glucoside, ascorbyl glucoside, l-ascorbic acid 2-o-alpha-glucoside, l-ascorbicacid2-glucosideShow MoreShow Less
PubChem CID 54693473
ChEBI CHEBI:81685
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2H-furan-5-one
SMILES C(C1C(C(C(C(O1)OC2=C(C(OC2=O)C(CO)O)O)O)O)O)O

Specifications

Melting Point 158°C to 163°C
Color White
Quantity 1 g
Solubility Information Soluble in water. (879g/L) at 25°C.
Formula Weight 338.26
Percent Purity 99.5%
Physical Form Powder
Chemical Name or Material 2-O-alpha-D-Glucopyranosyl-L-ascorbic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY08H072 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ66601.03,J66601.06
Lot NumberW02A017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66601.03,J66601.06
Lot NumberX24C032 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66601.03,J66601.06
Lot NumberY12A053 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ66601.03,J66601.06
Lot NumberS08C007 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ66601.03,J66601.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.