missing translation for 'onlineSavingsMsg'
Learn More
2-Thiohydantoin, 98%, Thermo Scientific Chemicals Product Code.: 15720809

2-Thiohydantoin, 98%, Thermo Scientific Chemicals

Product Code. 15720809
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 15720809

Brand: Thermo Scientific Acros 138761000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503-87-7
Molecular Formula C3H4N2OS
Molecular Weight (g/mol) 116.14
MDL Number MFCD00005277
InChI Key UGWULZWUXSCWPX-UHFFFAOYSA-N
Synonym 2-thiohydantoin, thiohydantoin, 4-imidazolidinone, 2-thioxo, 2-thioxoimidazolidin-4-one, 2-thioguidanthion, hydantoin, 2-thio, usaf be-25, 2-thioxo-4-imidazolidinone, 2-thioxo-1,3-diazolidin-4-one, glycine thiohydantoinShow MoreShow Less
PubChem CID 1274030
IUPAC Name 2-sulfanylideneimidazolidin-4-one
SMILES O=C1CNC(=S)N1

Specifications

Melting Point 229.0°C to 231.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 g
Beilstein 24,26
Solubility Information Solubility in water: slightly.
Formula Weight 116.14
Percent Purity 98%
Chemical Name or Material 2-Thiohydantoin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0444002 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number138761000,138760250
Lot NumberA0403265 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number138761000,138760050,138760250
Lot NumberA0418076 Certificate TypeCertificate of Analysis Date26/09/2022 Catalog Number138761000,138760050,138760250
Lot NumberA0444002 Certificate TypeCertificate of Analysis Date21/07/2022 Catalog Number138761000,138760050,138760250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Thiohydantoin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.