missing translation for 'onlineSavingsMsg'
Learn More
2-Vinylpyridine, 97%, stabilized, Thermo Scientific Chemicals Product Code.: 10295530

2-Vinylpyridine, 97%, stabilized, Thermo Scientific Chemicals

Product Code. 10295530
250 mL, Glass bottle
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10295530

Brand: Thermo Scientific Acros 158002500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-69-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006355
InChI Key KGIGUEBEKRSTEW-UHFFFAOYSA-N
Synonym 2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238Show MoreShow Less
PubChem CID 7521
IUPAC Name 2-ethenylpyridine
SMILES C=CC1=CC=CC=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -50°C
Color Brown
Density 0.9983g/mL
Boiling Point 159°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CH=CH2
Refractive Index 1.5490 to 1.551
Quantity 250 mL
Beilstein 20, 256
Specific Gravity 0.9983
Solubility Information Solubility in water: 2.5g/100 mL in water (20°C). Other solubilities: soluble in dilute acids,soluble in hydrocarbons,alcohol,ketones and,esters,soluble in methanol,ethanol,dioxane
Stabilizer 0.1% tert.-butylcatechol
Viscosity 1.2 mPa.s (20°C)
Formula Weight 105.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Vinylpyridine, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435555 Certificate TypeCertificate of Analysis Date24/04/2023 Catalog Number158000050,158000010,158002500
Lot NumberA0361098 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog Number158000050,158000010,158002500
Lot NumberA0409105 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog Number158000050,158000010,158002500
Lot NumberA0402283 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158000050,158000010,158002500
Lot NumberA0400866 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158000050,158000010,158002500
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Vinylpyridine, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Vinylpyridine, 97%, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.