missing translation for 'onlineSavingsMsg'
Learn More
2-Vinylpyridine, 97%, stabilized, Thermo Scientific Chemicals Product Code.: 10589633

2-Vinylpyridine, 97%, stabilized, Thermo Scientific Chemicals

Product Code. 10589633
1 L, Glass bottle
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10589633

Brand: Thermo Scientific Acros 158000010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-69-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006355
InChI Key KGIGUEBEKRSTEW-UHFFFAOYSA-N
Synonym 2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238Show MoreShow Less
PubChem CID 7521
IUPAC Name 2-ethenylpyridine
SMILES C=CC1=CC=CC=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -50°C
Color Brown
Density 0.9983g/mL
Boiling Point 159°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CH=CH2
Refractive Index 1.5490 to 1.551
Quantity 1 L
Beilstein 20, 256
Specific Gravity 0.9983
Solubility Information Solubility in water: 2.5g/100 mL in water (20°C). Other solubilities: soluble in dilute acids,soluble in hydrocarbons,alcohol,ketones and,esters,soluble in methanol,ethanol,dioxane
Stabilizer 0.1% tert.-butylcatechol
Viscosity 1.2 mPa.s (20°C)
Formula Weight 105.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Vinylpyridine, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435555 Certificate TypeCertificate of Analysis Date24/04/2023 Catalog Number158000050,158000010,158002500
Lot NumberA0361098 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog Number158000050,158000010,158002500
Lot NumberA0409105 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog Number158000050,158000010,158002500
Lot NumberA0402283 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158000050,158000010,158002500
Lot NumberA0400866 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158000050,158000010,158002500
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Vinylpyridine, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
2-Vinylpyridine, 97%, stabilized, Thermo Scientific Chemicals >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis