missing translation for 'onlineSavingsMsg'
Learn More
3-(2-Furyl)-1H-pyrazol-5-amine, 95+%, Thermo Scientific™ Product Code.: 10783953

3-(2-Furyl)-1H-pyrazol-5-amine, 95+%, Thermo Scientific™

Product Code. 10783953
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10783953

Brand: Thermo Scientific Maybridge KM00168DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 96799-02-9
Molecular Formula C7H7N3O
Molecular Weight (g/mol) 149.153
InChI Key XJNZHICOWTVWOX-UHFFFAOYSA-N
Synonym 3-furan-2-yl-1h-pyrazol-5-amine, 3-2-furyl-1h-pyrazol-5-amine, 5-furan-2-yl-1h-pyrazol-3-amine, 5-amino-3-2-furyl pyrazole, 3-amino-5-2-furyl pyrazole, 5-2-furyl-1h-pyrazol-3-amine, 3-amino-5-2-furyl-1h-pyrazole, 5-2-furanyl-1h-pyrazol-3-amine, 5-amino-3-fur-2-yl-1h-pyrazole, 5-furan-2-yl-2h-pyrazol-3-amineShow MoreShow Less
PubChem CID 523183
IUPAC Name 5-(furan-2-yl)-1H-pyrazol-3-amine
SMILES C1=COC(=C1)C2=CC(=NN2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 149.15
Percent Purity 95%
Chemical Name or Material 3-(2-Furyl)-1H-pyrazol-5-amine
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(2-Furyl)-1H-pyrazol-5-amine, 95+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.