Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass Stopcocks
Brand:  Lenz Laborglasinstrumente™ 02221402
 View more versions of this product
Product Code. 10622571
  • £251.00 / Pack of 10
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Stopcock
Plug size: NS 14.5
DIN 12541-1
2.5 mm
Borosilicate Glass
Solid Glass plug, capillary side arms
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass Stopcocks > Bore Size: 2.5mm; Includes Solid Glass plug, capillary side arms; Size: NS 14.5