missing translation for 'onlineSavingsMsg'
Learn More
3-Aminobenzeneboronic acid, 98%, Thermo Scientific Chemicals Product Code.: 15429687

3-Aminobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15429687
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15429687

Brand: Thermo Scientific Alfa Aesar H52909.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

Solubility
Soluble in water (partly miscible).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30418-59-8
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD00007755
InChI Key JMZFEHDNIAQMNB-UHFFFAOYSA-N
Synonym 3-aminobenzeneboronic acid, 3-aminophenyl boronic acid, m-aminophenylboronic acid, 3-aminophenyl boranediol, m-aminophenyl boronic acid, 3-amino phenylboronic acid, chembl20852, boronic acid, 3-aminophenyl, m-aminophenyl metaboric acidShow MoreShow Less
PubChem CID 92269
IUPAC Name (3-aminophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)N)(O)O

Specifications

Melting Point 138°C to 143°C
Quantity 25 g
Solubility Information Soluble in water (partly miscible).
Formula Weight 136.95
Percent Purity 98%
Chemical Name or Material 3-Aminobenzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW17K003 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog NumberH52909.03,H52909.14,H52909.06
Lot NumberU05K033 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberH52909.03,H52909.14,H52909.06
Lot NumberY15I011 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog NumberH52909.03,H52909.14,H52909.06
Lot NumberS24I052 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberH52909.03,H52909.14,H52909.06
Lot NumberU05G021 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52909.03,H52909.14,H52909.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Aminobenzeneboronic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt