missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Bromo-4,5-dihydro-isoxazol-5-ylmethyl)carbamic acid tert-butyl ester, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
Specifications
Specifications
CAS | 109770-82-3 |
Assay Percent Range | 97% |
Molecular Formula | C9H15BrN2O3 |
MDL Number | MFCD08741480 |
Quantity | 10 g |
Synonym | tert-butyl 3-bromo-4,5-dihydroisoxazol-5-yl methyl carbamate, 3-bromo-4,5-dihydro-isoxazol-5-ylmethyl carbamic acid tert-butyl ester, n-boc-3-bromo-2-isoxazoline-5-methylamine, tert-butyl n-3-bromo-4,5-dihydro-1,2-oxazol-5-yl methyl carbamate, 3-bromo-4,5-dihydro-isoxazol-5-ylmethyl-carbamic acid tert-butyl ester, acmc-20a3tu, tert-butyl 3-bromo-4,5-dihydroisoxazol-5-yl methylcarbamate, 5-n-tert-butoxycarbonyl-aminomethyl-3-bromo-4,5-dihydroisoxazole, tert-butyl 3-bromo-4,5-dihydro-1,2-oxazol-5-yl methyl carbamate |
InChI Key | AHAHZEKELWWMSJ-UHFFFAOYNA-N |
SMILES | CC(C)(C)OC(=O)NCC1CC(Br)=NO1 |
IUPAC Name | tert-butyl N-[(3-bromo-4,5-dihydro-1,2-oxazol-5-yl)methyl]carbamate |
Molecular Weight (g/mol) | 279.13 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(3-Bromo-4,5-dihydro-isoxazol-5-ylmethyl)carbamic acid tert-butyl ester, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction