missing translation for 'onlineSavingsMsg'
Learn More
3-Iodo-4-nitroanisole, 97%, Thermo Scientific Chemicals Product Code.: 10611762

3-Iodo-4-nitroanisole, 97%, Thermo Scientific Chemicals

Product Code. 10611762
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10611762

Brand: Thermo Scientific Acros 326320050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214279-40-0
Molecular Formula C7H6INO3
Molecular Weight (g/mol) 279.03
MDL Number MFCD01320682
InChI Key NLTAHTYBZZSCIT-UHFFFAOYSA-N
Synonym 3-iodo-4-nitroanisole, 4-nitro-3-iodoanisole, benzene, 2-iodo-4-methoxy-1-nitro, 1-nitro-2-iodo-4-methoxybenzene, 3-iodo-4-nitrophenyl methyl ether, benzene,2-iodo-4-methoxy-1-nitroShow MoreShow Less
PubChem CID 2733456
IUPAC Name 2-iodo-4-methoxy-1-nitrobenzene
SMILES COC1=CC=C(C(I)=C1)[N+]([O-])=O

Specifications

Melting Point 68.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 279.03
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Iodo-4-nitroanisole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0357168 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number326320250,326320050
Lot NumberA0357168 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number326320250,326320050
compliance-icons compliance-icons
Product Identifier
  • 3-Iodo-4-nitroanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Iodo-4-nitroanisole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.