Learn More
TubeWriter 300 - Standard Rack Side Print for Vacutainer Tubes
Standard rack for directly printing on the side of vacutainer tubes
Brand:  TubeWriter PS-BDVACUTAINER-300
Product Code. 30067436
  • £942.00 / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Standard printing rack designed for 300
  • Prints and labels directly on the side of vacutainer tubes
  • Rack ensures correct alignment and holds tubes securely
  • No time-consuming sticky labels or handwriting required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Specifications
Specifications
Labeling Rack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TubeWriter 300 - Standard Rack Side Print for Vacutainer Tubes > Standard Rack Side Print for Vacutainer Tubes