Learn More
Gibco™ Cell Dissociation Buffer, enzyme-free, PBS
Animal origin-free
Brand: Gibco™ 13151014
25.64 GBP valid until 2025-12-31
Use promo code "24335" to get your promotional price.
Description
Gibco Cell Dissociation Buffer is a membrane-filtered, isotonic, and enzyme-free solution of salts, chelating agents, and cell-conditioning agents in calcium-free and magnesium-free phosphate-buffered saline (PBS). Gibco Cell Dissociation Buffer is suitable for the gentle dissociation of mammalian cells for studies such as ligand binding, flow cytometry, and immunohistochemistry, that require intact cell surface proteins. Gibco Cell Dissociation Buffer works well with lightly adherent cell lines, such as HeLa and NIH 3T3, but is not recommended for the routine passaging of strongly adherent cells.
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature

Specifications
EDTA | |
HeLa, NIH 3T3 | |
Cell Culture Dissociation Reagent | |
EDTA | |
Room Temperature Stable |
Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 24 months from date of manufacture |
|
Animal Origin-free | |
Sterile-filtered | |
100 mL | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467354 | Certificate of Analysis | 31/01/2025 | 131510250,131510010,131510025,13151014 |
2572391 | Certificate of Analysis | 29/01/2025 | 13151014 |
2572392 | Certificate of Analysis | 29/01/2025 | 13151014 |
2944208 | Certificate of Analysis | 25/01/2025 | 13151014 |
A0446664 | Certificate of Analysis | 09/05/2024 | 131510250,131510010,131510025,13151014 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.