missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Accutase
Novel trypsin replacement that exhibits both protease and collagenolytic activities.
Brand: MP Biomedicals™ 091000449
89.27 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Novel trypsin replacement that exhibits both protease and collagenolytic activities for cell detachment
- Effective in detaching primary fibroblasts, endothelial cells, neurons, tumor cell lines and insect cells
- Maintains most cell surface antigens

Specifications
-20°C | |
Sterile | |
100 mL |
Cell Culture Dissociation Reagent | |
6.8 to 7.8 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Accutase > Accutase, 100 mL
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction