missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, indicator grade Product Code.: 10552764

Thermo Scientific Chemicals 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, indicator grade

Product Code. 10552764
1 kg
Click to view available options
Quantity:
25 g
100 g
1 kg
This item is not returnable. View return policy

Product Code. 10552764

Brand: Thermo Scientific Chemicals 401260010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-73-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.38
MDL Number MFCD00038130
InChI Key MLVYOYVMOZFHIU-UHFFFAOYSA-M
Synonym orange iv, tropaeolin oo, orangeiv, acid orange 5, diphenylamine orange, tropaeolin, unii-vr8q3r288w, tropeolin oo, orange n, solar orange ivShow MoreShow Less
PubChem CID 23668839
IUPAC Name sodium;4-[(4-anilinophenyl)diazenyl]benzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Color Ochre to Orange
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 kg
Merck Index 15, 9955
Formula Weight 375.38
Grade Extra Pure
Physical Form Powder
Chemical Name or Material 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459550 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number401260250,401260010,401261000
Lot NumberA0399958 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number401260250,401260010,401261000
Lot NumberA0385553 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number401260250,401260010,401261000
Lot NumberA0409066 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number401260250,401260010,401261000
Lot NumberA0448208 Certificate TypeCertificate of Analysis Date08/12/2022 Catalog Number401260250,401260010,401261000
compliance-icons
Product Identifier
  • 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, indicator grade
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, indicator grade >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.