missing translation for 'onlineSavingsMsg'
Learn More
4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™, Thermo Scientific Chemicals Product Code.: 11995681

4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11995681
1 L, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy

Product Code. 11995681

Brand: Thermo Scientific Acros 431790010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5124-30-1
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.35
MDL Number MFCD00015405
InChI Key KORSJDCBLAPZEQ-UHFFFAOYSA-N
Synonym hydrogenated mdi, hylene w, bis 4-isocyanatocyclohexyl methane, 4,4'-diisocyanatodicyclohexylmethane, methylene bis 4-cyclohexylisocyanate, dicyclohexylmethane 4,4'-diisocyanate, dicyclohexylmethane-4,4'-diisocyanate, methylene bis-4-cyclohexylisocyanate, nacconate h 12, cyclohexane, 1,1'-methylenebis 4-isocyanatoShow MoreShow Less
PubChem CID 21202
ChEBI CHEBI:53216
IUPAC Name 1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
SMILES O=C=NC1CCC(CC2CCC(CC2)N=C=O)CC1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 19.0°C to 23.0°C
Color Colorless
Density 1.0660g/mL
Boiling Point 112°C to 114°C (1010.0 hPa)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Quantity 1 L
Specific Gravity 1.066
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone
Formula Weight 262.35
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 4, 4'-Methylenebis(cyclohexyl isocyanate)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0433035 Certificate TypeCertificate of Analysis Date30/04/2024 Catalog Number431790010,431791000
Lot NumberA0447436 Certificate TypeCertificate of Analysis Date14/12/2023 Catalog Number431790010,431791000
Lot NumberA0406063 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number431790010,431791000
Lot NumberA0422275 Certificate TypeCertificate of Analysis Date22/09/2020 Catalog Number431790010,431791000
compliance-icons compliance-icons
Product Identifier
  • 4,4'-Methylenebis(cyclohexyl isocyanate)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.