missing translation for 'onlineSavingsMsg'
Learn More
4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals Product Code.: 15435939

4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals

Product Code. 15435939
500 mg
Click to view available options
Quantity:
100 mg
500 mg
1 g
This item is not returnable. View return policy

Product Code. 15435939

Brand: Thermo Scientific Alfa Aesar J63763.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Analog of genistein and an agonist at estrogen recpetors

Analog of genistein and an agonist at estrogen recpetors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00016954
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyroneShow MoreShow Less
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

Melting Point 328°C to 332°C (decomposition)
Color White to Yellow
Quantity 500 mg
Beilstein 231523
Merck Index 14,2801
Formula Weight 254.24
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 4',7-Dihydroxyisoflavone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY13K045 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ63763.03,J63763.MC,J63763.ME
Lot NumberR01K029 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog NumberJ63763.03,J63763.MC,J63763.ME
Lot NumberY04I035 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ63763.03,J63763.MC,J63763.ME
Lot NumberZ15H030 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ63763.03,J63763.MC,J63763.ME
Lot NumberU04H012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63763.03,J63763.MC,J63763.ME

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.