missing translation for 'onlineSavingsMsg'
Learn More
4-Amino-2-chloro-5-fluoropyrimidine, 98%, Thermo Scientific Chemicals Product Code.: 10244864

4-Amino-2-chloro-5-fluoropyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 10244864
1 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10244864

Brand: Thermo Scientific Acros 399980010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155-10-2
Molecular Formula C4H3ClFN3
Molecular Weight (g/mol) 147.54
MDL Number MFCD00057344
InChI Key SLQAJWTZUXJPNY-UHFFFAOYSA-N
Synonym 4-amino-2-chloro-5-fluoropyrimidine, 2-chloro-5-fluoro-pyrimidin-4-ylamine, 2-chloro-4-amino-5-fluoropyrimidine, 2-chloro-5-fluoro-4-aminopyrimidine, 2-chloro-5-fluoropyrimidin-4-ylamine, 2-chloro-5-fluoropyrimidine-4-ylamine, 4-pyrimidinamine, 2-chloro-5-fluoro, 4-pyrimidinamine, 2-chloro-5-fluoro-9ci, pubchem6930, 2-chlor-4-amino-5-fluorpyrimidinShow MoreShow Less
PubChem CID 254372
IUPAC Name 2-chloro-5-fluoropyrimidin-4-amine
SMILES C1=C(C(=NC(=N1)Cl)N)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 193°C to 195°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 147.54
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chloro-5-fluoropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-2-chloro-5-fluoropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Amino-2-chloro-5-fluoropyrimidine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.