missing translation for 'onlineSavingsMsg'
Learn More
4-Benzyloxyphenylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals Product Code.: 11965981

4-Benzyloxyphenylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11965981
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy

Product Code. 11965981

Brand: Thermo Scientific Acros 436221000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 120186-59-6, 109-99-9
CAS Min % 19.48, 74.96
CAS Max % 25.04, 80.52
Color Colorless to Brown
Density 1.0330g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C13H11BrMgO
Linear Formula C6H5CH2OC6H4MgBr
MDL Number MFCD09039130
Quantity 100 mL
Specific Gravity 1.033
Synonym 4-benzyloxyphenylmagnesium bromide solution, magnesium bromide 4-benzyloxy benzen-1-ide 1/1/1, magnesium 2+ ion 4-benzyloxy benzen-1-ide bromide, grignard reagent
InChI Key NMSSOXCJYITCNG-UHFFFAOYSA-M
SMILES Br[Mg]C1=CC=C(OCC2=CC=CC=C2)C=C1
IUPAC Name magnesium;phenylmethoxybenzene;bromide
Molecular Weight (g/mol) 287.44
PubChem CID 11098170
Formula Weight 287.43
Physical Form Liquid
Chemical Name or Material 4-Benzyloxyphenylmagnesium bromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465648 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog Number436221000
Lot NumberA0459157 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number436221000
Lot NumberA0431450 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number436221000
Lot NumberA0370843 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number436221000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Benzyloxyphenylmagnesium bromide, 0.8M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Benzyloxyphenylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.