missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-3-phenyl-1H-pyrazol-5-amine, ≥95%, Thermo Scientific™ Product Code.: 10377064

4-Bromo-3-phenyl-1H-pyrazol-5-amine, ≥95%, Thermo Scientific™

Product Code. 10377064
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10377064

Brand: Thermo Scientific Maybridge KM00411EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2845-78-5
Molecular Formula C9H8BrN3
Molecular Weight (g/mol) 238.09
MDL Number MFCD00082666
InChI Key QTNVXMOPTHGCII-UHFFFAOYSA-N
Synonym 4-bromo-3-phenyl-1h-pyrazol-5-amine, 3-amino-4-bromo-5-phenylpyrazole, 4-bromo-5-phenyl-2h-pyrazol-3-amine, 3-amino-4-bromo-5-phenyl-1h-pyrazole, 1h-pyrazol-3-amine,4-bromo-5-phenyl, acmc-20amn3, maybridge1_004579, 4-bromo-3-phenylpyrazole-5-ylamine, 4-bromo-5-phenylpyrazole-3-ylamine, 4-bromo-3-phenyl-1h-pyrazole-5-amineShow MoreShow Less
PubChem CID 594319
IUPAC Name 4-bromo-5-phenyl-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1Br)C1=CC=CC=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 238.09
Percent Purity 95%
Chemical Name or Material 4-Bromo-3-phenyl-1H-pyrazol-5-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-3-phenyl-1H-pyrazol-5-amine, ≥95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.