missing translation for 'onlineSavingsMsg'
Learn More
4-Chloropyridine hydrochloride, 98+%, Thermo Scientific Chemicals Product Code.: 11469160

4-Chloropyridine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11469160
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11469160

Brand: Thermo Scientific Alfa Aesar A15628.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7379-35-3
Molecular Formula C5H5Cl2N
Molecular Weight (g/mol) 150.00
MDL Number MFCD00012829
InChI Key XGAFCCUNHIMIRV-UHFFFAOYSA-N
Synonym 4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochlorideShow MoreShow Less
PubChem CID 81852
IUPAC Name 4-chloropyridine;hydrochloride
SMILES [H+].[Cl-].ClC1=CC=NC=C1

Specifications

Melting Point ∼210°C (sublimation)
pH 2
Flash Point 198°C (388°F)
Odor Weak
Quantity 5 g
Beilstein 3684904
Solubility Information Soluble in water.
Formula Weight 150.01
Percent Purity ≥98%
Chemical Name or Material 4-Chloropyridine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10240173 Certificate TypeCertificate of Analysis Date14/04/2022 Catalog NumberA15628.22,A15628.06,A15628.14
Lot Number10156436 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15628.22,A15628.06,A15628.14
Lot Number10137599 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15628.22,A15628.06,A15628.14
Lot Number10190701 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15628.22,A15628.06,A15628.14
Lot Number10160792 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15628.22,A15628.06,A15628.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloropyridine hydrochloride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.