missing translation for 'onlineSavingsMsg'
Learn More
4-Dimethylamino-1-naphthoic acid, 98%, Thermo Scientific Chemicals Product Code.: 11312117

4-Dimethylamino-1-naphthoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11312117
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11312117

Brand: Thermo Scientific Alfa Aesar L07944.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78062-03-0
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD00051797
InChI Key LZQKNRRVTOJUHQ-UHFFFAOYSA-N
Synonym 4-dimethylamino-1-naphthoic acid, 4-dimethylamino naphthalene-1-carboxylic acid, 4-dimethylamino-1-naphthalenecarboxylic acid, 4-dimethylamino naphthalic acid, 4-dimethylamino-1-naphthoicacid, 4-dimethylamino-1-naphthoic acid #, 4-dimethylaminonaphthalenecarboxylic acid, 4-dimethylamino naphthalenecarboxylic acid, 4-dimethylamino-naphthalene-1-carboxylic acidShow MoreShow Less
PubChem CID 522695
IUPAC Name 4-(dimethylamino)naphthalene-1-carboxylic acid
SMILES CN(C)C1=CC=C(C2=CC=CC=C21)C(=O)O

Specifications

Melting Point ∽160°C (decomposition)
Quantity 1 g
Beilstein 2723206
Formula Weight 215.25
Percent Purity 98%
Chemical Name or Material 4-Dimethylamino-1-naphthoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10112553 Certificate TypeCertificate of Analysis Date22/05/2023 Catalog NumberL07944.03,L07944.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Dimethylamino-1-naphthoic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.