missing translation for 'onlineSavingsMsg'
Learn More
4-Methyltetrahydropyran, stabilized with BHT, Thermo Scientific Chemicals Product Code.: 17291302
compliance-icons compliance-icons

4-Methyltetrahydropyran, stabilized with BHT, Thermo Scientific Chemicals

Product Code. 17291302
1 L
Click to view available options
Quantity:
100 mL
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 17291302

Brand: Thermo Scientific Acros 467320010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4717-96-8
Molecular Weight (g/mol) 100.16

Specifications

Density 0.8570 g/mL
Boiling Point 105.0°C
Flash Point 6.5°C
Molecular Formula C6H12O
Quantity 1 L
Specific Gravity 0.857
Solubility Information Solubility aq. soln.: 19.2 g/L (20°C).
Viscosity 0.78 mPa.s (20°C)
Formula Weight 100.16
Chemical Name or Material 4-Methyltetrahydropyran

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0420160 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number467320010,467321000,467320025
Lot NumberA0459356 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number467320010,467321000,467320025
compliance-icons compliance-icons
Product Identifier
  • 4-Methyltetrahydropyran, stabilized with BHT
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydro-4-methyl-2H-pyran

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methyltetrahydropyran, stabilized with BHT, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.