missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Okadaic acid sodium salt, 98%, high purity
Click to view available options
Quantity:
0.1 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 209266-80-8 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Molecular Formula | C44H67NaO13 |
MDL Number | MFCD00210208 |
Quantity | 0.1 mg |
Synonym | okadaic acid sodium salt, okadaic acid sodium salt from prorocentrum concavum, okadaic acid sodium salt from prorocentrum concavum hplc , film |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0412307 | Certificate of Analysis | 25/03/2024 | 328400001 |
Safety and Handling

Product Identifier
- Okadaic acid sodium salt (high purity)
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Okadaic acid sodium salt, 98%, high purity >
Spot an opportunity for improvement?Share a Content Correction