missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals Product Code.: 11483494

4-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11483494
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11483494

Brand: Thermo Scientific Alfa Aesar A11194.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store at 2-8°C. Incompatible with water, heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-28-7
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
MDL Number MFCD00007306
InChI Key GFNKTLQTQSALEJ-UHFFFAOYSA-N
Synonym 4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanateShow MoreShow Less
PubChem CID 66012
IUPAC Name 1-isocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=O)[N+](=O)[O-]

Specifications

Melting Point 56°C to 59°C
Boiling Point 137°C to 138°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3335
Beilstein 389516
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 164.12
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl isocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10239999 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11194.03,A11194.14,A11194.06
Lot Number10231223 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11194.03,A11194.14,A11194.06
Lot Number10196180 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11194.03,A11194.14,A11194.06
Lot Number10144639 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11194.03,A11194.14,A11194.06
Lot NumberL07Q020 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11194.03,A11194.14,A11194.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.