missing translation for 'onlineSavingsMsg'
Learn More
5,5-Dimethyl-1,3-dioxan-2-one, tech., Thermo Scientific Chemicals Product Code.: 11372696

5,5-Dimethyl-1,3-dioxan-2-one, tech., Thermo Scientific Chemicals

Product Code. 11372696
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11372696

Brand: Thermo Scientific Alfa Aesar L08767.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3592-12-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00014650
InChI Key JRFXQKZEGILCCO-UHFFFAOYSA-N
Synonym neopentylene carbonate, 2,2-dimethyltrimethylene carbonate, 5,5-dimethyl-1,3-diox-2-one, acmc-1adx1, 3-nitro-5-chlorobenzenesulfonamide, 5,5-dimethyltrimethylene carbonate, 5,5-dimethyl-1,3-dioxane-2-one, 5,5-dimethyl-2-oxo-1,3-dioxane, 1,3-dioxan-2-one,5,5-dimethyl, 5,5-dimethyl-1,3 dioxan-2-oneShow MoreShow Less
PubChem CID 137984
IUPAC Name 5,5-dimethyl-1,3-dioxan-2-one
SMILES CC1(COC(=O)OC1)C

Specifications

Melting Point 105°C to 114°C
Quantity 1 g
Beilstein 109055
Formula Weight 130.14
Grade Technical
Chemical Name or Material 5,5-Dimethyl-1,3-dioxan-2-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG27L21 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08767.06,L08767.03
Lot Number10214843 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08767.06,L08767.03
Lot Number10229273 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL08767.06,L08767.03
Lot NumberG15N30 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL08767.06,L08767.03
Lot Number10215800 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL08767.06,L08767.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,5-Dimethyl-1,3-dioxan-2-one, tech., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.