missing translation for 'onlineSavingsMsg'
Learn More
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%, Thermo Scientific Chemicals Product Code.: 11421057

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%, Thermo Scientific Chemicals

Product Code. 11421057
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11421057

Brand: Thermo Scientific Alfa Aesar B24066.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4-disulfonic acid monosodium salt hydrate is used in the accurate determination of sub microgram levels of serum iron. (i.e.. reagent for the spectrophotometric determination of Fe).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4″-disulfonic acid monosodium salt hydrate is used in the accurate determination of sub microgram levels of serum iron. (i.e.. reagent for the spectrophotometric determination of Fe).

Solubility
Soluble in water (partly).

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-85-3
Molecular Formula C20H15N4NaO7S2
Molecular Weight (g/mol) 510.471
MDL Number MFCD00150794
InChI Key MBOKSYFCYXIJRZ-UHFFFAOYSA-M
Synonym ferrozine mono-sodium salt hydrate, pdt disulfonate monosodium salt hydrate, 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine-4,4-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p inverted exclamation marka-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid monosodium salt hydrate, sodium 4-3-pyridin-2-yl-6-4-sulfophenyl-1,2,4-triazin-5-yl benzenesulfonate hydrateShow MoreShow Less
PubChem CID 71311205
IUPAC Name sodium;4-[3-pyridin-2-yl-6-(4-sulfophenyl)-1,2,4-triazin-5-yl]benzenesulfonate;hydrate
SMILES C1=CC=NC(=C1)C2=NC(=C(N=N2)C3=CC=C(C=C3)S(=O)(=O)O)C4=CC=C(C=C4)S(=O)(=O)[O-].O.[Na+]

Specifications

Quantity 1 g
Beilstein 5222087
Solubility Information Soluble in water (partly).
Formula Weight 492.47 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulfonic acid monosodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244368 Certificate TypeCertificate of Analysis Date28/10/2022 Catalog NumberB24066.06,B24066.03,B24066.14
Lot Number10213171 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24066.06,B24066.03,B24066.14
Lot Number10237240 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24066.06,B24066.03,B24066.14
Lot Number10211466 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24066.06,B24066.03,B24066.14
Lot Number10107657 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24066.06,B24066.03,B24066.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.