missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | C6H5C(O)NHCH(CO2H)CH2CH2CH2NHC(NH)NH2 |
Quantity | 1 g |
Formula Weight | 278.31 |
Percent Purity | 97% |
Chemical Name or Material | 5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction