missing translation for 'onlineSavingsMsg'
Learn More
5-Bromo-2,3-dihydrobenzo[b]furan-7-boronic acid, 97%, Thermo Scientific™ Product Code.: 10320453

5-Bromo-2,3-dihydrobenzo[b]furan-7-boronic acid, 97%, Thermo Scientific™

Product Code. 10320453
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10320453

Brand: Thermo Scientific Maybridge MO07179CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-72-5
Molecular Formula C8H8BBrO3
Molecular Weight (g/mol) 242.86
MDL Number MFCD06200893
InChI Key BSSWVOKZPQTKGW-UHFFFAOYSA-N
Synonym 5-bromo-2,3-dihydrobenzofuran-7-yl boronic acid, 5-bromo-2,3-dihydrobenzo b furan-7-boronic acid, 5-bromo-2,3-dihydro-1-benzofuran-7-yl boronic acid, acmc-1b7su, 5-bromo-2,3-dihydrobenzofuran-7-yl boronicacid, boronic acid, b-5-bromo-2,3-dihydro-7-benzofuranylShow MoreShow Less
PubChem CID 2794805
IUPAC Name (5-bromo-2,3-dihydro-1-benzofuran-7-yl)boronic acid
SMILES OB(O)C1=CC(Br)=CC2=C1OCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 242.86
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 3-dihydrobenzo[b]furan-7-boronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-2,3-dihydrobenzo[b]furan-7-boronic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.