missing translation for 'onlineSavingsMsg'
Learn More
5-Bromopyridine-2-carboxaldehyde, 99%, Thermo Scientific Chemicals Product Code.: 11479270

5-Bromopyridine-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 11479270
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11479270

Brand: Thermo Scientific Alfa Aesar H50154.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Bromopyridine-2-carboxaldehyde is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyridine-2-carboxaldehyde is used as organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31181-90-5
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD04112535
InChI Key ZQVLPMNLLKGGIU-UHFFFAOYSA-N
Synonym 5-bromopicolinaldehyde, 5-bromo-2-formylpyridine, 5-bromo-2-pyridinecarboxaldehyde, 5-bromopyridine-2-carboxaldehyde, 5-bromo-pyridine-2-carbaldehyde, 5-bromo-2-pyridinecarbaldehyde, 2-pyridinecarboxaldehyde, 5-bromo, 5-bromo-2-pyridine-carbaldehyde, pubchem5059, 5-bromo picolinaldehydeShow MoreShow Less
PubChem CID 9877562
IUPAC Name 5-bromopyridine-2-carbaldehyde
SMILES C1=CC(=NC=C1Br)C=O

Specifications

Melting Point 94°C to 96°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 186.01
Percent Purity 99%
Chemical Name or Material 5-Bromopyridine-2-carboxaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY11I011 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberH50154.03,H50154.06
Lot NumberM26H027 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50154.03,H50154.06
Lot NumberA31S012 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50154.03,H50154.06
Lot NumberI19S029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50154.03,H50154.06
Lot Number10169679 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50154.03,H50154.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromopyridine-2-carboxaldehyde, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.