missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-2-methoxyphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 188132-02-7, 109-99-9 |
CAS Min % | 9.68, 85.49 |
CAS Max % | 14.51, 90.32 |
Color | Yellow to Brown |
Density | 0.9480g/mL |
Flash Point | −17°C |
Packaging | AcroSeal™ Glass Bottle |
Molecular Formula | C7H6BrFMgO |
Linear Formula | F(OCH3)C6H3MgBr |
MDL Number | MFCD01311481 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Fluoro-2-methoxyphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™ >
Spot an opportunity for improvement?Share a Content Correction