missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 5mL Polystyrene Immuno-Tubes

Use for determining hormone levels.

Brand:  Greiner Bio-One 115061

 View more versions of this product

Product Code. 13077573

  • £126.54 / Pack of 2000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Round bottom
  • Dia. x Height: 12 x 75mm
  • Available with either medium or high binding capacities
  • With or without star at bottom
  • Star at the bottom of the tubes serve to increase the surface and thus make it possible to bind amounts of antigens or antibodies
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Determining hormone levels
Immuno Tube
75 mm
5 mL
Polystyrene
High-binding
12 mm
Without Star Bottom
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Greiner Bio-One 5mL Polystyrene Immuno-Tubes > High binding; Without star at the bottom of tube

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.