missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro-imidazo 2,1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro, acmc-1c5af, 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonylchloride, 6-chloro-5-imidazo 2,1-b thiazolesulfonyl chloride, 6-chloro-imidazo 2, 1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride,6-chloro |
Formula Weight | 257.12 |
Percent Purity | 97% |
Chemical Name or Material | 6-Chloroimidazo[2, 1-b][1, 3]thiazole-5-sulfonyl chloride |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
6-Chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction