Learn More
Thermo Scientific™ 6-Place Fixed-Angle Highconic™ Rotor Adapters
Use these rotor adapters to connect with round or conical-bottom centrifuge tubes and microtubes tubes up to 50mL.
Brand:  Thermo Scientific™ 76002905
 View more versions of this product
Product Code. 10107881
  • £82.98 / Each
Explore more special offers
Requires : 50mL conical adapter 75-007-556 requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
Description
Description
Use these 6-Place Fixed-Angle Highconic™ Rotor Adapters to connect with centrifuge tubes up to 50mL. The adapters accommodate round or conical-bottom tubes and microtubes and work with HiGhConic™ fixed angle rotors.
The adapters accommodate round- or conical-bottom tubes and microtubes.
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microtube
HiGhConic™ fixed angle rotor
1.5 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ 6-Place Fixed-Angle Highconic™ Rotor Adapters > Adapter; 4 x 1.5mL; 24 Tubes