missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 850 Homogenizer Package
GREENER_CHOICE

Fisher Scientific™ 850 Homogenizer Package includes everything needed to disrupt, emulsify or homogenize a wide range of samples in volumes ranging from 200μL to 5L. Includes motor, stand assembly and 3 stainless steel probes.

Brand:  Fisherbrand™ 15515819

5104.65 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 15515819

  • £5495.00 / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Warning: Stainles Steel bearings cannot be autoclaved.

This item is not returnable. View return policy

Description

Description

  • Includes 10 x 115mm, 20 x 195mm, and 30 x 195mm stainless steel generator probes
  • Powerful 850 watt motor
  • Integrated digital speed control displays speed and run time
  • Variable speed control from 5,000 to 25,000rpm
  • Can be used with plastic generator probes (adapter required for use)
  • Processing range from 0.2mL to 5L
  • Sound level: <72 db.

Certifications

CE

Services

Two years

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Homogenizer
115 V
8.5 in.
Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
56 oz.
5000 to 25,000 rpm
115 V, 50/60 Hz
21.5 cm
  • 850 motor
  • 10x115 mm probe
  • 20x195 mm probe
  • 30x195 mm probe
  • 850 motor drive, tool kit, mounting stand clamp and instruction manual
  • Motor Stand
1.6 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    8

    Hours

    3

    Minutes

    23

    Seconds

    18
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ 850 Homogenizer Package > 850 Homogenizer Package

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.