missing translation for 'onlineSavingsMsg'
Learn More

uvex 95798 Insole For Uvex 1 Sport - Width 11

Comfortable climatic insole with antistatic properties for Uvex 1 Sport

£17.30

Specifications

Color Black
For Use With (Equipment) Uvex 1 Sport
Material Microvelour
Disposable Single-use
Gender Unisex
View More Specs

Products 16
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17248971
View Documents
uvex
9579835
35
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17258971
View Documents
uvex
9579836
36
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17268971
View Documents
uvex
9579837
37
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17278971
View Documents
uvex
9579838
38
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17288971
View Documents
uvex
9579839
39
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17298971
View Documents
uvex
9579840
40
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17208981
View Documents
uvex
9579841
41
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17218981
View Documents
uvex
9579842
42
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17228981
View Documents
uvex
9579843
43
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17238981
View Documents
uvex
9579844
44
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17248981
View Documents
uvex
9579845
45
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17258981
View Documents
uvex
9579846
46
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17268981
View Documents
uvex
9579847
47
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17278981
View Documents
uvex
9579848
48
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17288981
View Documents
uvex
9579849
49
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
17298981
View Documents
uvex
9579851
51
£17.30
1 pair
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
Description

Description

  • Full-surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No. 9579.8
  • Sizes: 35 to 52
Specifications

Specifications

Black
Microvelour
Unisex
Yes
Insoles
Uvex 1 Sport
Single-use
Standard Width
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
uvex 95798 Insole For Uvex 1 Sport - Width 11

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.