Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi-Purpose Tube Rotator
Optional accessories to enhance the Fisherbrand™Multi-Purpose Tube Rotator
Brand:  Fisherbrand™ 88861126
 View more versions of this product
Product Code. 15504100
  • £127.00 / Each
Estimated Shipment: 24-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Multi-purpose tube rotator offers flexibility with multiple carousel options
  • Tube rotator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
64 place drum tube carousel for 18.5mm dia (15-20mL culture tubes)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Multi-Purpose Tube Rotator > 64 place drum tube carousel for 18.5mm dia (15-20mL culture tubes)