missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, for HPLC, Fisher Chemical™ Product Code.: 10754361

Acetonitrile, for HPLC, Fisher Chemical™

Product Code. 10754361
1 L, Amber Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
This item is not returnable. View return policy

Product Code. 10754361

Brand: Fisher Chemical A/0626/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053, Amber glass bottle, Liquid, 6342, CC#N, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -46°C
Color Colorless
Boiling Point 81°C
Quantity 1 L
Packaging Amber Glass Bottle
Vapor Pressure 97mbar at 20°C
Viscosity 0.36cp at 20°C

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2528119 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberA/0626/27SS,A/0626/99W,A/0626/21,A/0626/PB17X,A/0626/10RSS,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17,A/0626/30RSS
Lot Number2528118 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberA/0626/27SS,A/0626/99W,A/0626/21,A/0626/PB17X,A/0626/10RSS,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17,A/0626/30RSS
Lot Number2527244 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberA/0626/99W,A/0626/10RSS,A/0626/30RSS,A/0626/27SS,A/0626/21,A/0626/PB17,A/0626/PB17X,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17
Lot Number2527243 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberA/0626/99W,A/0626/10RSS,A/0626/30RSS,A/0626/27SS,A/0626/21,A/0626/PB17,A/0626/PB17X,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17
Lot Number2526614 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberA/0626/99W,A/0626/10RSS,A/0626/30RSS,A/0626/27SS,A/0626/21,A/0626/PB17,A/0626/PB17X,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Acetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, for HPLC, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.