Learn More
Cytiva Activated Calf Thymus DNA
High-quality activated calf thymus DNA
Brand:  Cytiva 27-4575-01
Product Code. 10349874
  • £293.00 / Each
Explore more special offers
Description
Description
  • Treated with DNase I
For preparing substrate used in the study of DNA polymerases
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Liquid
Viral and eukaryote
25 Units
Activated Calf Thymus DNA
25 μg/mL
Study of DNA Polymerase
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Activated Calf Thymus DNA > Activated calf thymus DNA