Learn More
Eppendorf™ Adapters For Round Bucket in Rotor A-4-38
Wide variety of adapters for all blood collection tubes and cell culture tube types
Brand:  Eppendorf™ 5702724009
168.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11572215
  • £168.67 / Pack of 2
Explore more special offers
Description
Description
  • Adapter for Rotor A-4-38
  • For centrifugation of blood collection tubes or conical tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
9 to 15 mL
17.5 mm
2 Pack
Monovette
Round bucket in Rotor A-4-38
16
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Adapters For Round Bucket in Rotor A-4-38 > Bore Size: 17.5mm; For Use With: Round bucket in Rotor A-4-38