missing translation for 'onlineSavingsMsg'
Learn More
Adipic dihydrazide, 98%, Thermo Scientific Chemicals Product Code.: 10531651

Adipic dihydrazide, 98%, Thermo Scientific Chemicals

Product Code. 10531651
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10531651

Brand: Thermo Scientific Acros 203310250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-93-8
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00007614
InChI Key IBVAQQYNSHJXBV-UHFFFAOYSA-N
Synonym adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5mShow MoreShow Less
PubChem CID 66117
IUPAC Name hexanedihydrazide
SMILES NNC(=O)CCCCC(=O)NN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 178.0°C to 182.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula H2NNHCO(CH2)4CONHNH2
Quantity 25 g
Beilstein 02, I, 277
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetic acid,slightly soluble in acetone,insoluble in ethanol,ether and benzene
Formula Weight 174.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Adipic dihydrazide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461242 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number203310250,203311000,203315000
Lot NumberA0432316 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number203310250,203311000,203315000
Lot NumberA0391910 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number203310250,203311000,203315000
Lot NumberA0411513 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number203310250,203311000,203315000
Lot NumberA0446249 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog Number203310250,203311000,203315000
compliance-icons compliance-icons
Product Identifier
  • Adipic dihydrazide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Adipic dihydrazide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.