Learn More
Motic™ AE31E Trinocular Microscope
Use this microscope for routine live cell inspection. With an easy and flexible set up, it is ideal for University, routine-clinical and lab environments. Motic™ AE31E Trinocular Microscopes offer scale research functions, such as fluorescence and an externally mounted and centrable 30W/6V halogen Koehler illumination.
Brand:  Motic™ 1100100200576
Product Code. 15363087
  • £6060.00 / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Widefield high eyepoint eyepieces WFPL10X/22mm with diopter adjustment on both eyepieces, with rubber eyecups
  • Centering telescope
  • Side facing quintuple nosepiece
  • CCIS Plan Achromatic objective PL4X
  • CCIS Plan Achromatic Phase objective PL PH10X, LWD PL PH20X, LWD PL PH40X
  • Plain stage, glass and metal inserts
  • ELWD N.A. 0.30 condenser
  • Phase slider, centrable PH1, brightfield, PH3
  • Koehler illumination quartz halogen 6V/30W with external lamp house and intensity control
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Motic™ AE31E Trinocular Microscope > AE31E Trinocular Microscope