missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BB15 CO2 Incubator

The Thermo Scientific BB15 CO2 incubator provides everything you need for the good growth of cell and tissue cultures.

Brand:  Thermo Scientific™ 51023122

5274.07 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.


 View more versions of this product

Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 12846716

  • £8718.38 / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ BB15 CO2 incubator provides everything you need for the good growth of cell and tissue cultures. The chamber has been designed for a less contamination-prone surface (up to 50% less surface) than other incubators.

  • Precise temperature and CO2 control for optimal growth conditions
  • Powerful humidification system for short recovery times
  • Solid copper interior for permanent contamination control (optional)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5.3 cu. ft.
Copper
CO2 Incubator
Water reservoir
470 x 607 x 530 mm / 18.5 x 23.9 x 20.9 in.
230 V, 50/60 Hz
BB15 150L Single CO2 Incubator
TC Sensor
General Purpose
637 x 867 x 766 mm/ 25.1 x 34.1 x 30.2 in.
70kg or 154 lbs
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ BB15 CO2 Incubator >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.