Learn More
Applied Biosystems™ ExoSAP-IT™ For PCR Product Clean-Up
Designed for simple, quick PCR cleanup for downstream applications, such as DNA sequencing or Single Nucleotide Polymorphism (SNP) analysis. Single-step enzymatic cleanup of PCR products that eliminates unincorporated primers and dNTPs.
Brand:  Applied Biosystems™ 78202.4X.1.ML
 View more versions of this product
Product Code. 15593677
  • £1604.00 / 2000 reactions
Explore more special offers
Description
Description
  • Two hydrolytic enzymes, Exonuclease I and Shrimp Alkaline Phosphatase (SAP), together in a specially formulated buffer
  • 30-minute protocol
TRUSTED_SUSTAINABILITY
Specifications
Specifications
-20°C
2000
4 x 1 mL
Quick PCR clean-up for downstream applications; Eliminates unincorporated primers and dNTPs
Single-step enzymatic PCR Cleanup Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ ExoSAP-IT™ For PCR Product Clean-Up > 2000 reactions