Learn More
AIM-100, MedChemExpress
MedChemExpress AIM-100 is a potent and selective Ack1 inhibitor with an IC50 of 21.58 nM. AIM-100 also inhibits Tyr267 phosphorylation. AIM-100 does not inhibits other kinases including PI3-kinase and AKT subfamily members. AIM-100 has an anticancer effect.
Brand: Medchem Express HY-15290
Description
AIM-100 is a potent and selective Ack1 inhibitor with an IC50 of 21.58 nM. AIM-100 also inhibits Tyr267 phosphorylation. AIM-100 does not inhibits other kinases including PI3-kinase and AKT subfamily members. AIM-100 has an anticancer effect.Specifications
AIM-100 | |
Light Yellow | |
C23H21N3O2 | |
DMSO : 50 mg/mL (134.61 mM; Need ultrasonic) | |
371.43 | |
99.92% | |
Solid |
873305-35-2 | |
Research | |
100 mg | |
C1(C2=C(C3=CC=CC=C3)OC4=C2C(NC[C@@H]5CCCO5)=NC=N4)=CC=CC=C1 | |
371.43 | |
Research | |
Cancer-Kinase/protease |
Product Suggestions
Customers who viewed this item also viewed.