missing translation for 'onlineSavingsMsg'
Learn More

R&L Enterprises™ Reaction Vessel Clamp

Designed to support Quickfit reaction vessels. R&L Enterprises™ Reaction Vessel Clamps have glass fibre covered jaws opening up to 180mm maximum.

Brand:  R&L Enterprises™ 2124

Product Code. 10360295

  • £172.21 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Each clamp provided with two extension rods, one for use with vessels up to 5L capacity and the other to 20L
  • Rods can be secured to the main body using socket screws, a wrench for which is provided
  • Rod diameter is 9.5mm
Specifications

Specifications

1 Ea.
Alloy, Zinc-Plated Steel
Heat Resistant Covered
Reaction Vessel Clamp
For use with up to 5/20L vessels
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&L Enterprises™ Reaction Vessel Clamp > For Use With: For use with up to 5/20L vessels

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.