Learn More
Ansell AlphaTec™ 4000 Cape Hood
Engineered to provide an exceptional barrier against a wide range of organic and inorganic chemicals and biological agents.
Brand:  Ansell GR40W-00507-00
926.16 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17170855
  • £1300.00 / Pack of 100
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Specifications
One Size
Green
Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes
Multi-layer Barrier Laminate Fabric
Welded
G40W507
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ansell AlphaTec™ 4000 Cape Hood > Style: G40W507