Learn More
Ansell AlphaTec™ 38-628 Butyl/Viton Immersion Gloves
AlphaTec™ Gloves 38-628 offer a combination of butyl and viton layer of 0.7 mm for a maximum protection against hazardous chemicals. Provide the best resistance to the most aggressive chemicals without compromising dexterity or comfort. Offer superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids.
£298.00 - £309.92
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17814851
|
Ansell
38628070 |
7 |
£298.00
1 pair |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
17824851
|
Ansell
38628110 |
11 |
£298.00
1 pair |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
17182122
|
Ansell
38628080 |
8 |
£309.92
1 pair |
In Stock
Log in to see stock. |
||||||
Description
- AlphaTec™ 38-628 offers superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids
- The glove is very flexible and provides excellent dexterity
- Designed for comfort, with a natural, curved ergonomic shape and soft feel, offering easy donning and good grip
- The absence of latex proteins means there's no risk of latex allergies
Specifications
Yes |