missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 38-628 Butyl/Viton Immersion Gloves

AlphaTec™ Gloves 38-628 offer a combination of butyl and viton layer of 0.7 mm for a maximum protection against hazardous chemicals. Provide the best resistance to the most aggressive chemicals without compromising dexterity or comfort. Offer superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids.

Brand:  Ansell

124.76 GBP valid until 2025-04-18
Use promo code "24095" to get your promotional price.


 View more versions of this product

Product Code. 17182122

  • £309.92 / 1 pair
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Provides the best resistance to the most aggressive chemicals, without compromising dexterity or comfort
  • Flexible and excellent dexterity, designed for comfort  with a natural, curved ergonomic shape and soft feel
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black
Viton/Butyl Polymer
8
Yes
Industrial Gloves
Rolled
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use code 24095 to receive this promotion.
    Promo Code: 24095

    Expires2025-04-15

    Days

    10

    Hours

    13

    Minutes

    46

    Seconds

    52
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 17/03/2025 and 15/04/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.

    Terms & Conditions Learn More
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell AlphaTec™ 38-628 Butyl/Viton Immersion Gloves > AhaTec Protection Gloves; Color: Black; Material : Viton / Butyl Polymer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.