missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ RNA Micro Scale Kit

PureLink™ technology combines guanidine-isothiocyanate lysis with the speed, purity, and ease-of-use of silica-membrane purification.

Marca:  Invitrogen™ 12183016

Código de producto. 10184903

  • £348.00 / 50 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore más ofertas especiales

For Research Use Only. All usage must comply with product instructions.
 
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

A simple and reliable technology for everyday RNA purification
PureLink™ technology combines guanidine-isothiocyanate lysis with the speed, purity, and ease-of-use of silica-membrane purification. The safe and easy procedure can be completed in typically less than 15 minutes with >85% recovery from the first elution. The kit can be used for clean-up of purified RNA from enzymatic reactions, desalting of RNA samples after phenol extraction or ethanol precipitation, and concentration of RNA. Purify RNA from up to 10 mg of tissue or 105 cells. Clean up and concentrate RNA amounts ranging from pg to 100 μg in volumes as small as 10 μL. Purified RNA is ready for use in downstream applications such as RT-PCR, qRT-PCR, northern blots, cDNA library construction, and nuclease protection assays.

A simple and rapid procedure
Homogenize your sample in lysis buffer, add 70% ethanol, and mix. Deposit the resulting solution into a PureLink Micro kit column with collection tube, and centrifuge for one minute. Nucleic acids bind to the PureLink silica membrane in the column, and after subsequent washes and on-column digestion with DNase, purified RNA can be eluted with RNase-free water.

  • Effective: Expect excellent RNA recovery (up to 100 μg total RNA) from small samples
  • Convenient and safe: The extraction procedure uses no phenol/chloroform, CsCl gradients, or precipitation with either LiCl or ethanol
  • Fast: Obtain high-quality total RNA, typically in just 15 minutes
  • Reliable: Obtain ready-to-use RNA suitable for any downstream application
TRUSTED_SUSTAINABILITY
Especificaciones
Show More
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

One moment while we fetch your results.
Certificados

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3007769 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12183016
Lot Number3020862 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12183016
Lot Number3106298 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12183016
Lot Number2471941 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12183016
Lot Number2500117 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12183016
Ofertas especiales

Ofertas especiales

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Invitrogen™ PureLink™ RNA Micro Scale Kit > 50 Preps

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado

For Research Use Only. Not for use in diagnostic procedures.